2-(Furfurylthio)ethylamine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-{[(2-Aminoethyl)thio]methyl}furan
OR42233-1G
OR42233-250MG
OR42233-500MG
OR42233-5G
APOSOR42233-1GEA
1490
CZK
APOSOR42233-1G
APOSOR42233-250MG
APOSOR42233-500MG
APOSOR42233-5G
2-(Furfurylthio)ethylamine
2-(Furfurylthio)ethylamine
Vzorec:
C₇H₁₁NOS Hustota: 1,13 g/cm³ (25 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD01075029 CAS číslo: 36415-21-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...