Tisk…

3-Chloro-4-(ethylcarbamoyl)phenylboronic acid 97%

Dodavatel: Apollo Scientific

Synonyms: 3-Chloro-4-(ethylcarbamoyl)benzeneboronic acid, (3-Chloro-4-(ethylcarbamoyl)phenyl)boronic acid

OR4218-10G OR4218-1G OR4218-5G
APOSOR4218-10GEA 29790 CZK
APOSOR4218-10G APOSOR4218-1G APOSOR4218-5G
3-Chloro-4-(ethylcarbamoyl)phenylboronic acid 97%
3-Chloro-4-(ethylcarbamoyl)phenylboronic acid
Vzorec: C₉H₁₁BClNO₃
MW: 227,45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07363767
CAS číslo: 850589-40-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR