2-Methyl-4-pyridineboronic acid pinacol ester
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-Methylpyridine-4-boronic acid Pinacol ester, 2-(2-Methyl-4-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
OR42154-1G
OR42154-25G
OR42154-5G
APOSOR42154-1GEA
744
CZK
APOSOR42154-1G
APOSOR42154-25G
APOSOR42154-5G
2-Methyl-4-pyridineboronic acid pinacol ester
2-Methyl-4-pyridineboronic acid pinacol ester
Vzorec:
C₁₂H₁₈BNO₂ Bod tání: 66…71 °C Storage Temperature: Freezer |
MDL Number:
MFCD08061590 CAS číslo: 660867-80-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...