Tisk…

Ethyl-4-(4-chlorophenyl)-2,4-dioxobutanoate

Dodavatel: Apollo Scientific
Varování

OR42147-1G OR42147-250MG OR42147-5G
APOSOR42147-1GEA 4570 CZK
APOSOR42147-1G APOSOR42147-250MG APOSOR42147-5G
Ethyl-4-(4-chlorophenyl)-2,4-dioxobutanoate
Ethyl-4-(4-chlorophenyl)-2,4-dioxobutanoate
Vzorec: C₁₂H₁₁ClO₄
Storage Temperature: Ambient
MDL Number: MFCD01935972
CAS číslo: 5814-38-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR