2-Indanylacetic acid 99%
Dodavatel: Apollo Scientific
Synonyms:
2-(2,3-Dihydro-1H-inden-2-yl)acetic acid
OR42111-1G
OR42111-5G
APOSOR42111-1GEA
3710
CZK
APOSOR42111-1G
APOSOR42111-5G
2-Indanylacetic acid 99%
2-Indanylacetic acid
Vzorec:
C₁₁H₁₂O₂ MW: 176,22 g/mol Bod tání: 91…93 °C Bod vzplanutí: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00082653 CAS číslo: 37868-26-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...