Tisk…

3-Isobutylamino-propan-1-ol 95%

Dodavatel: Apollo Scientific

OR42060-1G OR42060-250MG OR42060-500MG
APOSOR42060-1GEA 11380 CZK
APOSOR42060-1G APOSOR42060-250MG APOSOR42060-500MG
3-Isobutylamino-propan-1-ol 95%
3-Isobutylamino-propan-1-ol
Vzorec: C₇H₁₇NO
Storage Temperature: Ambient
MDL Number: MFCD03446756
CAS číslo: 285124-45-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR