3-(1-Naphthyl)propionic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR4167-1G
OR4167-5G
OR4167-25G
APOSOR4167-1GEA
1240
CZK
APOSOR4167-1G
APOSOR4167-5G
APOSOR4167-25G
3-(1-Naphthyl)propionic acid
3-(1-Naphthyl)propionic acid
Vzorec:
C₁₃H₁₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00033058 CAS číslo: 3243-42-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...