4-(2-Cyanophenyl)morpholine 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Morpholinobenzonitrile, 2-(Morpholin-4-yl)benzonitrile
OR4129-1G
OR4129-5G
APOSOR4129-1GEA
3540
CZK
APOSOR4129-1G
APOSOR4129-5G
4-(2-Cyanophenyl)morpholine 98%
4-(2-Cyanophenyl)morpholine
Vzorec:
C₁₁H₁₂N₂O MW: 188,2258 g/mol Bod tání: 48…50 °C Storage Temperature: Ambient |
MDL Number:
MFCD01313390 CAS číslo: 204078-32-0 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...