Bromocyclobutane 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Cyclobutyl bromide
OR41168-25G
OR41168-5G
APOSOR41168-25GEA
1240
CZK
APOSOR41168-25G
APOSOR41168-5G
Bromocyclobutane 98%
Bromocyclobutane
Vzorec:
C₄H₇Br MW: 135 g/mol Bod varu: 107…108 °C Hustota: 1,434 g/cm³ (25 °C) Bod vzplanutí: 22 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00001317 CAS číslo: 4399-47-7 EINECS: 224-530-9 UN: 1993 ADR: 3,II |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...