Tisk…

Ethyl-6-bromoindole-2-carboxylate 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: Ethyl 6-bromoindole-2-carboxylate

OR41055-5G
APOSOR41055-5GEA 3110 CZK
APOSOR41055-5G
Ethyl-6-bromoindole-2-carboxylate 98%
Ethyl-6-bromoindole-2-carboxylate
Vzorec: C₁₁H₁₀BrNO₂
MW: 268,11 g/mol
Bod tání: 178…188 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03094942
CAS číslo: 103858-53-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR