3,4'-Biphthalic anhydride
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2,3,3,4-biphenyl tetracarboxylic dianhydride, a-BPDA, Diphenyl-2,3,3',4'-tetracarboxylic Dianhydride
OR41022-1G
OR41022-5G
OR41022-25G
APOSOR41022-1GEA
622
CZK
APOSOR41022-1G
APOSOR41022-5G
APOSOR41022-25G
3,4'-Biphthalic anhydride
3,4'-Biphthalic anhydride
Vzorec:
C₁₆H₆O₆ MW: 294,22 g/mol Bod tání: 195…205 °C Storage Temperature: Ambient |
MDL Number:
MFCD09030654 CAS číslo: 36978-41-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...