Tisk…

3-(4-Bromo-1-methyl-1H-pyrazol-3-yl)aniline

Dodavatel: Apollo Scientific
Varování

OR4099-1G OR4099-250MG
APOSOR4099-1GEA 4960 CZK
APOSOR4099-1G APOSOR4099-250MG
3-(4-Bromo-1-methyl-1H-pyrazol-3-yl)aniline
3-(4-Bromo-1-methyl-1H-pyrazol-3-yl)aniline
Vzorec: C₁₀H₁₀BrN₃
Storage Temperature: Ambient
MDL Number: MFCD00067937
CAS číslo: 175201-77-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR