3-(Dipropylcarbamoyl)phenylboronic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(3-(Dipropylcarbamoyl)phenyl)boronic acid, 3-(N,N-Dipropylaminocarbonyl)benzeneboronic acid
OR4094-1G
OR4094-5G
APOSOR4094-1GEA
827
CZK
APOSOR4094-1G
APOSOR4094-5G
3-(Dipropylcarbamoyl)phenylboronic acid 98%
3-(Dipropylcarbamoyl)phenylboronic acid
Vzorec:
C₁₃H₂₀BNO₃ MW: 249.1100 g/mol Bod tání: 118…120 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06659889 CAS číslo: 850567-39-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...