(R)-1-Boc-3-Ethylpiperazine 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(R)-tert-Butyl 3-ethylpiperazine-1-carboxylate, (3R)-3-Ethylpiperazine, N1-BOC protected
OR40701-250MG
OR40701-1G
APOSOR40701-250MGEA
787
CZK
APOSOR40701-250MG
APOSOR40701-1G
(R)-1-Boc-3-Ethylpiperazine 95%
(R)-1-Boc-3-Ethylpiperazine
Vzorec:
C₁₁H₂₂N₂O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD07772111 CAS číslo: 438050-08-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...