Methyl-3-bromo-2-methylbenzoate 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Methyl 3-bromo-2-methylbenzoate
OR40654-100G
OR40654-25G
OR40654-5G
APOSOR40654-100GEA
3410
CZK
APOSOR40654-100G
APOSOR40654-25G
APOSOR40654-5G
Methyl-3-bromo-2-methylbenzoate 97%
Methyl-3-bromo-2-methylbenzoate
Vzorec:
C₉H₉BrO₂ MW: 229,0706 g/mol Bod tání: 29…31 °C Storage Temperature: Ambient |
MDL Number:
MFCD09031772 CAS číslo: 99548-54-6 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...