Tisk…

2-Bromo-3-thiophenecarboxylic acid 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Bromothiophene-3-carboxylic acid

OR40613-25G OR40613-5G OR40613-1G
APOSOR40613-25GEA 7250 CZK
APOSOR40613-25G APOSOR40613-5G APOSOR40613-1G
2-Bromo-3-thiophenecarboxylic acid 95%
2-Bromo-3-thiophenecarboxylic acid
Vzorec: C₅H₃BrO₂S
MW: 207,041 g/mol
Bod tání: 178…182 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05865213
CAS číslo: 24287-95-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR