Tisk…

4-Bromo-3-nitroaniline 98%

Dodavatel: Apollo Scientific
Varování

OR40599-1G OR40599-5G
APOSOR40599-1GEA 622 CZK
APOSOR40599-1G APOSOR40599-5G
4-Bromo-3-nitroaniline 98%
4-Bromo-3-nitroaniline
Vzorec: C₆H₅BrN₂O₂
MW: 217,02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01320678
CAS číslo: 53324-38-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR