3-Amino-5-methylpyridine-2-carboxylic acid amide
Dodavatel: Apollo Scientific
|
Varování
|
OR40508-1G
OR40508-250MG
OR40508-5G
APOSOR40508-1GEA
4140
CZK
APOSOR40508-1G
APOSOR40508-250MG
APOSOR40508-5G
3-Amino-5-methylpyridine-2-carboxylic acid amide
3-Amino-5-methylpyridine-2-carboxylic acid amide
Vzorec:
C₇H₉N₃O Storage Temperature: Ambient |
MDL Number:
MFCD15142820 CAS číslo: 1208081-47-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...