6-Chloro-3-pyridazinecarboxaldehyde
Dodavatel: Apollo Scientific
|
Varování
|
OR40466-250MG
OR40466-100MG
APOSOR40466-250MGEA
2900
CZK
APOSOR40466-250MG
APOSOR40466-100MG
6-Chloro-3-pyridazinecarboxaldehyde
6-Chloro-3-pyridazinecarboxaldehyde
Vzorec:
C₅H₃ClN₂O Storage Temperature: Ambient |
MDL Number:
MFCD10698041 CAS číslo: 303085-53-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...