2-Amino-5-methylphenylboronic acid pinacol ester
Dodavatel: Apollo Scientific
|
Varování
|
OR40461-1G
OR40461-250MG
APOSOR40461-1GEA
4930
CZK
APOSOR40461-1G
APOSOR40461-250MG
2-Amino-5-methylphenylboronic acid pinacol ester
2-Amino-5-methylphenylboronic acid pinacol ester
Vzorec:
C₁₃H₂₀BNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD06795664 CAS číslo: 948592-80-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...