Tisk…

4,6-Dimethoxypyrimidine-2-carboxylic acid ethyl ester 95+%

Dodavatel: Apollo Scientific
Varování

Synonyms: Ethyl 4,6-dimethoxypyrimidine-2-carboxylate

OR4044-1G OR4044-250MG OR4044-5G
APOSOR4044-1GEA 1780 CZK
APOSOR4044-1G APOSOR4044-250MG APOSOR4044-5G
4,6-Dimethoxypyrimidine-2-carboxylic acid ethyl ester 95+%
4,6-Dimethoxypyrimidine-2-carboxylic acid ethyl ester
Vzorec: C₉H₁₂N₂O₄
MW: 212,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03645366
CAS číslo: 128276-49-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR