Tisk…

2-(Hydroxymethyl)cyclohexanone

Dodavatel: Apollo Scientific

OR40419-1G OR40419-250MG OR40419-100MG OR40419-500MG OR40419-2.5G OR40419-5G
APOSOR40419-1GEA 21230 CZK
APOSOR40419-1G APOSOR40419-250MG APOSOR40419-100MG APOSOR40419-500MG APOSOR40419-2.5G APOSOR40419-5G
2-(Hydroxymethyl)cyclohexanone
2-(Hydroxymethyl)cyclohexanone
Vzorec: C₇H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09414735
CAS číslo: 5331-08-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR