Methoxyacetyl chloride 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR40393-100G
OR40393-25G
APOSOR40393-100GEA
3110
CZK
APOSOR40393-100G
APOSOR40393-25G
Methoxyacetyl chloride 95%
Methoxyacetyl chloride
Vzorec:
C₃H₅ClO₂ MW: 108.52 Bod varu: 112…113 °C Hustota: 1.187 g/cm³ (25 °C) Bod vzplanutí: 35 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000728 CAS číslo: 38870-89-2 EINECS: 254-169-2 UN: 2920 ADR: 8+3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...