Tisk…

[3-(Methylamino)-4-nitrophenyl]methanol 95+%

Dodavatel: Apollo Scientific

Synonyms: 5-(Hydroxymethyl)-N-methyl-2-nitroaniline

OR40346-1G OR40346-500MG
APOSOR40346-1GEA 15310 CZK
APOSOR40346-1G APOSOR40346-500MG
[3-(Methylamino)-4-nitrophenyl]methanol 95+%
[3-(Methylamino)-4-nitrophenyl]methanol
Vzorec: C₈H₁₀N₂O₃
MW: 182,18 g/mol
Bod tání: 90…93 °C
Storage Temperature: Ambient
MDL Number: MFCD16622776
CAS číslo: 1228182-77-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR