Methyl-2-[(4-aminobenzyl)amino]-5-nitrobenzenecarboxylate 95+%
Dodavatel: Apollo Scientific
Synonyms:
Methyl 2-[(4-aminobenzyl)amino]-5-nitrobenzoate
OR40345-1G
OR40345-500MG
APOSOR40345-1GEA
15310
CZK
APOSOR40345-1G
APOSOR40345-500MG
Methyl-2-[(4-aminobenzyl)amino]-5-nitrobenzenecarboxylate 95+%
Methyl-2-[(4-aminobenzyl)amino]-5-nitrobenzenecarboxylate
Vzorec:
C₁₅H₁₅N₃O₄ MW: 301,3 g/mol Bod tání: 166…168 °C Storage Temperature: Ambient |
MDL Number:
MFCD16622773 CAS číslo: 1228183-73-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...