(2-Chloropyridin-3-yl)acetonitrile 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR40338-10G
OR40338-25G
OR40338-5G
APOSOR40338-10GEA
4930
CZK
APOSOR40338-10G
APOSOR40338-25G
APOSOR40338-5G
(2-Chloropyridin-3-yl)acetonitrile 97%
(2-Chloropyridin-3-yl)acetonitrile
Vzorec:
C₇H₅ClN₂ MW: 152,5800 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09923767 CAS číslo: 101012-32-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...