3-Amino-5-chloropyridine 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Chloropyridin-3-amine
OR40335-1G
OR40335-25G
OR40335-5G
APOSOR40335-1GEA
2320
CZK
APOSOR40335-1G
APOSOR40335-25G
APOSOR40335-5G
3-Amino-5-chloropyridine 98%
3-Amino-5-chloropyridine
Vzorec:
C₅H₅ClN₂ MW: 128,56 g/mol Bod tání: 71…75 °C Storage Temperature: Ambient |
MDL Number:
MFCD03701386 CAS číslo: 22353-34-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...