2-Hydroxy-4-pyridineboronic acid pinacol ester
Dodavatel: Apollo Scientific
|
Varování
|
OR40294-1G
OR40294-250MG
OR40294-5G
APOSOR40294-1GEA
7770
CZK
APOSOR40294-1G
APOSOR40294-250MG
APOSOR40294-5G
2-Hydroxy-4-pyridineboronic acid pinacol ester
2-Hydroxy-4-pyridineboronic acid pinacol ester
Vzorec:
C₁₁H₁₆BNO₃ Storage Temperature: Ambient |
MDL Number:
MFCD11223287 CAS číslo: 1256358-90-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...