Tisk…

4-Biphenylcarbonyl chloride 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-Phenylbenzoyl chloride, 4-Biphenylcarbonyl chloride

OR4021-100G OR4021-250G
APOSOR4021-100GEA 7870 CZK
APOSOR4021-100G APOSOR4021-250G
4-Biphenylcarbonyl chloride 98%
4-Biphenylcarbonyl chloride
Vzorec: C₁₃H₉ClO
Bod varu: 160 °C/2mmHg
Bod tání: 110…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00000692
CAS číslo: 14002-51-8
EINECS: 237-804-8
UN: 3261
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR