Tisk…

Ethyl-3-amino-1-methyl-1H-pyrazole-4-carboxylate

Dodavatel: Apollo Scientific
Varování

Synonyms: Ethyl 3-amino-1-methyl-1H-pyrazole-4-carboxylate

OR40195-1G OR40195-5G OR40195-25G
APOSOR40195-1GEA 870 CZK
APOSOR40195-1G APOSOR40195-5G APOSOR40195-25G
Ethyl-3-amino-1-methyl-1H-pyrazole-4-carboxylate
Ethyl-3-amino-1-methyl-1H-pyrazole-4-carboxylate
Vzorec: C₇H₁₁N₃O₂
MW: 169.18 g/mol
Bod tání: 90…91 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00101268
CAS číslo: 21230-43-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR