Tisk…

4-Methyl-1H-indazole-5-boronic acid 96%

Dodavatel: Apollo Scientific
Varování

Synonyms: (4-Methyl-1H-indazol-5-yl)boronic acid

OR40151-500MG OR40151-1G OR40151-5G
APOSOR40151-500MGEA 1120 CZK
APOSOR40151-500MG APOSOR40151-1G APOSOR40151-5G
4-Methyl-1H-indazole-5-boronic acid 96%
4-Methyl-1H-indazole-5-boronic acid
Vzorec: C₈H₉BN₂O₂
MW: 175,9800 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12028644
CAS číslo: 1245816-09-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR