Tisk…

5-Methyl-1H-indazole-4-boronic acid 97%

Dodavatel: Apollo Scientific
Varování

OR40147-5G
APOSOR40147-5GEA 0 CZK
APOSOR40147-5G
5-Methyl-1H-indazole-4-boronic acid 97%
5-Methyl-1H-indazole-4-boronic acid
Vzorec: C₈H₉BN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD10001490
CAS číslo: 1245816-10-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR