5-Bromo-1,4-dimethyl-1H-indazole
Dodavatel: Apollo Scientific
|
Varování
|
OR40128-1G
OR40128-100MG
OR40128-250MG
APOSOR40128-1GEA
8930
CZK
APOSOR40128-1G
APOSOR40128-100MG
APOSOR40128-250MG
5-Bromo-1,4-dimethyl-1H-indazole
5-Bromo-1,4-dimethyl-1H-indazole
Vzorec:
C₉H₉BrN₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD12028624 CAS číslo: 1159511-80-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...