Tisk…

6-Bromo-4-methyl-1H-indazole

Dodavatel: Apollo Scientific
Varování

OR40121-100MG OR40121-1G
APOSOR40121-100MGEA 622 CZK
APOSOR40121-100MG APOSOR40121-1G
6-Bromo-4-methyl-1H-indazole
6-Bromo-4-methyl-1H-indazole
Vzorec: C₈H₇BrN₂
MW: 211,0600 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07781476
CAS číslo: 885520-98-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR