Methyl-5-iodo-2-methoxybenzoate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Methyl 5-iodo-2-methoxybenzoate, 5-Iodo-2-methoxybenzoic acid methyl ester
OR401043-10G
OR401043-1G
OR401043-5G
APOSOR401043-10GEA
3940
CZK
APOSOR401043-10G
APOSOR401043-1G
APOSOR401043-5G
Methyl-5-iodo-2-methoxybenzoate
Methyl-5-iodo-2-methoxybenzoate
Vzorec:
C₉H₉IO₃ MW: 292.07 Bod tání: 57…61 °C Storage Temperature: Ambient |
MDL Number:
MFCD02683862 CAS číslo: 40757-09-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...