3-Chloro-5-nitrobenzoic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR401023-1G
OR401023-25G
OR401023-5G
APOSOR401023-1GEA
870
CZK
APOSOR401023-1G
APOSOR401023-25G
APOSOR401023-5G
3-Chloro-5-nitrobenzoic acid 98%
3-Chloro-5-nitrobenzoic acid
Vzorec:
C₇H₄ClNO₄ MW: 201,57 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09263224 CAS číslo: 34662-36-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...