Tisk…

2-Chloro-6-nitrobenzaldehyde

Dodavatel: Apollo Scientific

OR401012-25G OR401012-5G
APOSOR401012-25GEA 13030 CZK
APOSOR401012-25G APOSOR401012-5G
2-Chloro-6-nitrobenzaldehyde
2-Chloro-6-nitrobenzaldehyde
Vzorec: C₇H₄ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD00007204
CAS číslo: 6361-22-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR