Tisk…

2-Chloro-6-methoxybenzaldehyde

Dodavatel: Apollo Scientific

OR400979-5G
APOSOR400979-5GEA 1700 CZK
APOSOR400979-5G
2-Chloro-6-methoxybenzaldehyde
2-Chloro-6-methoxybenzaldehyde
Vzorec: C₈H₇ClO₂
MW: 170,5900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08059233
CAS číslo: 29866-54-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR