Tisk…

5-Bromo-2-methylthiazole 96%

Dodavatel: Apollo Scientific
Varování

OR40092-1G OR40092-250MG
APOSOR40092-1GEA 2320 CZK
APOSOR40092-1G APOSOR40092-250MG
5-Bromo-2-methylthiazole 96%
5-Bromo-2-methylthiazole
Vzorec: C₄H₄BrNS
MW: 178,06 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09879074
CAS číslo: 57268-16-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR