1-[2-(Methoxycarbonyl)-4-nitrophenyl]-3-azetanecarboxylic acid 95+%
Dodavatel: Apollo Scientific
OR40091-1G
OR40091-500MG
APOSOR40091-1GEA
15310
CZK
APOSOR40091-1G
APOSOR40091-500MG
1-[2-(Methoxycarbonyl)-4-nitrophenyl]-3-azetanecarboxylic acid 95+%
1-[2-(Methoxycarbonyl)-4-nitrophenyl]-3-azetanecarboxylic acid
Vzorec:
C₁₂H₁₂N₂O₆ MW: 280.24 g/mol Bod tání: 178…180 °C Storage Temperature: Ambient |
MDL Number:
MFCD16140290 CAS číslo: 1234873-83-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...