Tisk…

5-Chloro-2-methoxy-3-nitrophenylbenzenecarboxylate 95+%

Dodavatel: Apollo Scientific

OR40090-1G OR40090-500MG
APOSOR40090-1GEA 25460 CZK
APOSOR40090-1G APOSOR40090-500MG
5-Chloro-2-methoxy-3-nitrophenylbenzenecarboxylate 95+%
5-Chloro-2-methoxy-3-nitrophenylbenzenecarboxylate
Vzorec: C₁₄H₁₀ClNO₅
Bod tání: 123…125 °C
Storage Temperature: Ambient
MDL Number: MFCD16140275
CAS číslo: 1221792-84-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR