Tisk…

5-Nitro-m-cresol 98%

Dodavatel: Apollo Scientific

OR400867-1G OR400867-5G
APOSOR400867-1GEA 5590 CZK
APOSOR400867-1G APOSOR400867-5G
5-Nitro-m-cresol 98%
5-Nitro-m-cresol
Vzorec: C₇H₇NO₃
Storage Temperature: Ambient
MDL Number: MFCD09038591
CAS číslo: 127818-58-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR