Tisk…

3-Bromo-4-iodobenzonitrile

Dodavatel: Apollo Scientific
Varování

OR400773-1G OR400773-5G OR400773-10G
APOSOR400773-1GEA 622 CZK
APOSOR400773-1G APOSOR400773-5G APOSOR400773-10G
3-Bromo-4-iodobenzonitrile
3-Bromo-4-iodobenzonitrile
Vzorec: C₇H₃BrIN
MW: 307,9100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09800817
CAS číslo: 1000577-94-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR