Tisk…

1-Bromo-3-chloro-2-iodobenzene 99%

Dodavatel: Apollo Scientific
Varování

OR400660-25G OR400660-5G
APOSOR400660-25GEA 4140 CZK
APOSOR400660-25G APOSOR400660-5G
1-Bromo-3-chloro-2-iodobenzene 99%
1-Bromo-3-chloro-2-iodobenzene
Vzorec: C₆H₃BrClI
MW: 317,348 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09907935
CAS číslo: 450412-28-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR