2-Amino-4-(methoxycarbonyl)benzoic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR40063-1G
OR40063-5G
OR40063-25G
APOSOR40063-1GEA
953
CZK
APOSOR40063-1G
APOSOR40063-5G
APOSOR40063-25G
2-Amino-4-(methoxycarbonyl)benzoic acid
2-Amino-4-(methoxycarbonyl)benzoic acid
Vzorec:
C₉H₉NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD08690068 CAS číslo: 85743-02-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...