4-Bromo-2-nitrobenzonitrile 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR400603-1G
OR400603-5G
OR400603-10G
APOSOR400603-1GEA
1620
CZK
APOSOR400603-1G
APOSOR400603-5G
APOSOR400603-10G
4-Bromo-2-nitrobenzonitrile 98%
4-Bromo-2-nitrobenzonitrile
Vzorec:
C₇H₃BrN₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00087225 CAS číslo: 79603-03-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...