2-Phenylethylboronic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR4006-1G
OR4006-5G
APOSOR4006-1GEA
622
CZK
APOSOR4006-1G
APOSOR4006-5G
2-Phenylethylboronic acid
2-Phenylethylboronic acid
Vzorec:
C₈H₁₁BO₂ Bod tání: 82…84 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631226 CAS číslo: 34420-17-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...