2,4,6-Trimethoxybenzyl alcohol 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR400585-1G
OR400585-5G
OR400585-10G
APOSOR400585-1GEA
1330
CZK
APOSOR400585-1G
APOSOR400585-5G
APOSOR400585-10G
2,4,6-Trimethoxybenzyl alcohol 95%
2,4,6-Trimethoxybenzyl alcohol
Vzorec:
C₁₀H₁₄O₄ Storage Temperature: Ambient |
MDL Number:
MFCD00060356 CAS číslo: 61040-78-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...