Tisk…

5-Methylpyrazin-2(1H)-one

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Hydroxy-5-methylpyrazine

OR40054-1G OR40054-5G
APOSOR40054-1GEA 5790 CZK
APOSOR40054-1G APOSOR40054-5G
5-Methylpyrazin-2(1H)-one
5-Methylpyrazin-2(1H)-one
Vzorec: C5H6N2O
MW: 110,1139 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01693937
CAS číslo: 20721-17-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR