5-Hydroxypyrazine-2-carboxylic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Oxo-4,5-dihydropyrazine-2-carboxylic acid
OR40051-100G
OR40051-25G
APOSOR40051-100GEA
5960
CZK
APOSOR40051-100G
APOSOR40051-25G
5-Hydroxypyrazine-2-carboxylic acid 98%
5-Hydroxypyrazine-2-carboxylic acid
Vzorec:
C5H4N2O3 MW: 140,1000 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11978170 CAS číslo: 34604-60-9 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...